Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127182.1 | NP_001120654.1 | 2 | 335 | ||
NM_018131.4 | NP_060601.3 | 2 | 457 | |||
XM_011539918.1 | XP_011538220.1 | 2 | 460 | |||
GenBank mRNA | AB091343.1 | - | 2 | 446 | ||
AK001402.1 | - | 1 | 57 | |||
AK223495.1 | - | 2 | 438 | |||
AK315536.1 | - | 2 | 454 | |||
AY788918.1 | - | 2 | 457 | |||
BC008947.2 | - | 2 | 437 | 3028566 | ||
BX640718.1 | - | 3 | 377 | |||
DA545074.1 | - | 2 | 335 |