Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018132.3 | NP_060602.2 | 7, 8 | 679 | ||
XM_005249205.1 | XP_005249262.1 | 7, 8 | 740 | |||
XM_011514723.2 | XP_011513025.2 | 6, 7 | 615 | |||
GenBank mRNA | AK001407.1 | - | 2, 3 | 229 | ||
AK293016.1 | - | 7, 8 | 684 | |||
BC016279.1 | - | 7, 8 | 669 | 3908895 |