Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WHAMM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080435.2 | NP_001073904.1 | 10 | 3786 | ||
XM_005272421.4 | XP_005272478.1 | 10 | 3402 | |||
XM_005272422.3 | XP_005272479.1 | 10 | 3149 | |||
XM_005272423.4 | XP_005272480.1 | 10 | 3388 | |||
XM_011521232.2 | XP_011519534.1 | 8 | 3598 | |||
GenBank mRNA | AK126887.1 | - | 10 | 3788 |