Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC163P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033296.1 | 5 | 876 | |||
NR_033297.1 | 5 | 919 | ||||
NR_033298.1 | 6 | 1004 | ||||
GenBank mRNA | AK054714.1 | - | 5 | 888 | ||
AK300867.1 | - | 6 | 894 | |||
BC036618.2 | - | 5 | 537 | |||
BC047421.1 | - | 6 | 973 | |||
BC101254.2 | - | 5 | 814 | |||
BC101255.2 | - | 5 | 771 | |||
BC101256.1 | - | 5 | 771 | |||
BC101257.1 | - | 6 | 899 | |||
BC113060.1 | - | 6 | 827 | |||
BC113061.1 | - | 6 | 894 |