Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TATDN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042552.2 | NP_001036017.1 | 8, 9 | 705 | ||
NM_001146169.1 | NP_001139641.1 | 9 | 726 | |||
NM_001146170.1 | NP_001139642.1 | 7, 8 | 642 | |||
NM_001146171.1 | NP_001139643.1 | 9 | 726 | |||
XM_011509202.2 | XP_011507504.1 | 7, 8 | 587 | |||
XM_017000328.1 | XP_016855817.1 | 8 | 658 | |||
XM_017000331.1 | XP_016855820.1 | 7, 8 | 584 | |||
GenBank mRNA | AK293129.1 | - | 9 | 726 | ||
AK304557.1 | - | 7, 8 | 600 | |||
BC048115.1 | - | 8, 9 | 616 | |||
BC113638.1 | - | 8, 9 | 649 | 8322690 | ||
BC117485.1 | - | 8, 9 | 649 | 40126036 | ||
BC143957.1 | - | 9 | 670 |