Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085447.1 | NP_001078916.1 | 8 | 1296 | ||
XM_011510589.1 | XP_011508891.1 | 8 | 1184 | |||
XM_011510590.1 | XP_011508892.1 | 8 | 1217 | |||
GenBank mRNA | AK301851.1 | - | 8 | 1264 | ||
BC015980.2 | - | 2 | 244 | |||
BC065905.1 | - | 3 | 264 | |||
BC117445.1 | - | 8 | 1239 | 40125996 |