Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018146.3 | NP_060616.1 | 1, 2 | 363 | ||
GenBank mRNA | AF177344.2 | - | 1, 2 | 419 | ||
AK001443.1 | - | 1, 2 | 339 | |||
AK222900.1 | - | 1, 2 | 339 | |||
AK225128.1 | - | 1, 2 | 320 | |||
BC011550.1 | - | 1, 2 | 325 | 4620063 | ||
BC050614.1 | - | 1, 2 | 321 | 5248052 |