Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080537.1 | NP_001074006.1 | 3 | 266 | ||
GenBank mRNA | AK126350.1 | - | 3 | 236 |