Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305395.1 | NP_001292324.1 | 4, 5 | 445 | ||
NM_001305396.1 | NP_001292325.1 | 4, 5 | 434 | |||
NM_001305397.1 | NP_001292326.1 | 4, 5 | 418 | |||
XM_017009029.1 | XP_016864518.1 | 4, 5 | 1632 | |||
XM_017009030.1 | XP_016864519.1 | 4, 5 | 1604 | |||
GenBank mRNA | BU561730.1 | - | 4, 5 | 381 | ||
HY000873.1 | - | 3, 4 | 357 | |||
HY001392.1 | - | 3, 4 | 341 |