Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF300P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026867.1 | 2, 3 | 346 | |||
GenBank mRNA | AK096536.1 | - | 2, 3 | 1501 | ||
AK097462.1 | - | 1, 2 | 204 | |||
BC117490.1 | - | 2, 3 | 308 | |||
BC126474.1 | - | 2, 3 | 308 |