Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128618.1 | NP_001122090.1 | 5 | 978 | ||
XM_011518225.2 | XP_011516527.1 | 4 | 822 | |||
GenBank mRNA | BC036255.1 | - | 5 | 978 | ||
BC044620.1 | - | 5 | 956 | |||
BC053956.1 | - | 5 | 956 | |||
BC130404.1 | - | 4 | 877 | 40146435 | ||
BC144273.1 | - | 4 | 876 | 9052793 |