Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other XRRA1 SIRNA Assays ›
Gene Symbol:
XRRA1
Gene Name:
X-ray radiation resistance associated 1
Gene Aliases:
-
Chromosome Location:
Chr.11: 74840910 - 74949187 on Build GRCh38
UniGene ID:
Hs.370145
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
264115
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: XRRA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001270380.1
NP_001257309.1
15
2944
NM_001270381.1
NP_001257310.1
16
3043
NM_182969.2
NP_892014.1
19
3311
XM_005273765.2
XP_005273822.1
19
3335
XM_005273770.2
XP_005273827.1
17
3115
XM_011544755.1
XP_011543057.1
19
3290
XM_011544756.1
XP_011543058.1
18
3209
XM_011544757.1
XP_011543059.1
18
3164
XM_011544758.1
XP_011543060.1
17
3164
XM_011544759.1
XP_011543061.1
17
3140
XM_011544761.1
XP_011543063.1
16
3092
XM_011544762.1
XP_011543064.1
16
3074
XM_011544763.1
XP_011543065.1
18
3229
XM_011544764.1
XP_011543066.1
16
2999
XM_011544765.1
XP_011543067.1
17
3049
XM_011544766.2
XP_011543068.1
19
3354
XM_011544767.1
XP_011543069.1
18
3256
XM_011544768.1
XP_011543070.1
17
3103
XM_017017221.1
XP_016872710.1
17
3125
XM_017017222.1
XP_016872711.1
17
3103
XM_017017223.1
XP_016872712.1
16
3058
XM_017017224.1
XP_016872713.1
15
2968
XM_017017225.1
XP_016872714.1
15
2893
XM_017017226.1
XP_016872715.1
18
3230
XM_017017227.1
XP_016872716.1
18
3206
XM_017017228.1
XP_016872717.1
16
3009
XM_017017229.1
XP_016872718.1
17
3059
XM_017017230.1
XP_016872719.1
16
2883
XM_017017231.1
XP_016872720.1
15
2969
XM_017017232.1
XP_016872721.1
13
2673
XM_017017233.1
XP_016872722.1
16
3063
XM_017017234.1
XP_016872723.1
15
2948
GenBank mRNA
AK074152.1
-
14
2641
BC037294.2
-
16
2976
BC040541.1
-
16
2976
CR749605.1
-
14
3415
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
response to X-ray
molecular_function
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-8tsm6:80/100.66.77.47:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline