Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGPEP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102612.2 | NP_001096082.2 | 4 | 644 | ||
NM_001167902.1 | NP_001161374.1 | 4 | 988 | |||
XM_011521254.1 | XP_011519556.1 | 5 | 683 | |||
XM_017021933.1 | XP_016877422.1 | 3 | 645 | |||
GenBank mRNA | AK058162.1 | - | 4 | 988 | ||
BC104743.1 | - | 4 | 550 | |||
BC104744.1 | - | 3 | 457 | |||
BC105686.1 | - | 3 | 457 |