Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGPEP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102612.2 | NP_001096082.2 | 5 | 1003 | ||
NM_001167902.1 | NP_001161374.1 | 5 | 1347 | |||
XM_011521254.1 | XP_011519556.1 | 6 | 1042 | |||
XM_017021933.1 | XP_016877422.1 | 4 | 1004 | |||
GenBank mRNA | AK058162.1 | - | 5 | 1348 | ||
BC104743.1 | - | 5 | 909 | |||
BC104744.1 | - | 4 | 817 | |||
BC105686.1 | - | 4 | 812 |