Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BICDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103175.1 | NP_001096645.1 | 6, 7 | 1148 | ||
XM_005255135.4 | XP_005255192.1 | 7, 8 | 1209 | |||
XM_011522391.1 | XP_011520693.1 | 6, 7 | 1397 | |||
GenBank mRNA | AL833717.1 | - | 4, 5 | 2354 | ||
AL833749.1 | - | 5, 6 | 2673 | |||
BC128602.1 | - | 6, 7 | 1148 | 40132962 |