Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SBNO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167856.1 | NP_001161328.1 | 27 | 3575 | ||
NM_018183.3 | NP_060653.3 | 27 | 3572 | |||
XM_005253572.3 | XP_005253629.1 | 28 | 3713 | |||
XM_005253573.4 | XP_005253630.1 | 29 | 4123 | |||
XM_005253575.3 | XP_005253632.1 | 28 | 3710 | |||
XM_005253576.3 | XP_005253633.1 | 29 | 3754 | |||
XM_006719473.3 | XP_006719536.1 | 30 | 3879 | |||
XM_006719474.3 | XP_006719537.1 | 29 | 3832 | |||
XM_011538533.2 | XP_011536835.1 | 30 | 3873 | |||
XM_011538534.2 | XP_011536836.1 | 27 | 3611 | |||
XM_011538535.2 | XP_011536837.1 | 27 | 3613 | |||
XM_017019556.1 | XP_016875045.1 | 28 | 3697 | |||
XM_017019557.1 | XP_016875046.1 | 28 | 3704 | |||
XM_017019558.1 | XP_016875047.1 | 29 | 3767 | |||
XM_017019559.1 | XP_016875048.1 | 28 | 3834 | |||
GenBank mRNA | AB014772.1 | - | 27 | 3572 | ||
AK001563.1 | - | 4 | 318 | |||
AK074256.1 | - | 11 | 1142 | |||
AK096864.1 | - | 10 | 945 | |||
BC133704.1 | - | 27 | 3575 | 40124902 |