Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHADL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138481.1 | NP_612490.1 | 5 | 2297 | ||
XM_005261370.3 | XP_005261427.1 | 5 | 2343 | |||
XM_011529932.2 | XP_011528234.1 | 4 | 2961 | |||
XM_011529933.2 | XP_011528235.1 | 4 | 2959 | |||
XM_011529934.2 | XP_011528236.1 | 4 | 2961 | |||
GenBank mRNA | BC012882.1 | - | 2 | 344 | ||
BC019839.1 | - | 3 | 1484 | |||
BC040188.1 | - | 5 | 2037 | |||
BC048421.1 | - | 3 | 1320 | |||
BC068590.1 | - | 5 | 2297 |