Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01315 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120595.1 | 2 | 1161 | |||
NR_120596.1 | 2 | 742 | ||||
NR_120597.1 | 2 | 399 |