Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HHAT SIRNA Assays ›
Gene Symbol:
HHAT
Gene Name:
hedgehog acyltransferase
Gene Aliases:
MART2, SKI1, Skn
Chromosome Location:
Chr.1: 210327873 - 210676298 on Build GRCh38
UniGene ID:
Hs.58650
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
26431
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HHAT
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001122834.3
NP_001116306.1
3
298
NM_001170564.2
NP_001164035.1
3
298
NM_001170580.2
NP_001164051.1
3
353
NM_001170587.2
NP_001164058.1
2
230
NM_001170588.2
NP_001164059.1
3
364
NM_018194.5
NP_060664.2
3
364
XM_006711441.3
XP_006711504.1
4
425
XM_011509737.2
XP_011508039.1
4
425
XM_011509738.2
XP_011508040.1
2
183
XM_011509739.2
XP_011508041.1
3
350
XM_011509740.2
XP_011508042.1
4
554
XM_011509741.2
XP_011508043.1
3
298
XM_011509742.2
XP_011508044.1
4
402
XM_011509746.2
XP_011508048.1
3
379
XM_017001728.1
XP_016857217.1
3
508
XM_017001729.1
XP_016857218.1
3
229
XM_017001732.1
XP_016857221.1
3
189
XM_017001735.1
XP_016857224.1
3
829
XM_017001736.1
XP_016857225.1
2
205
XM_017001737.1
XP_016857226.1
3
189
XM_017001738.1
XP_016857227.1
3
296
XM_017001739.1
XP_016857228.1
3
385
XM_017001740.1
XP_016857229.1
3
886
GenBank mRNA
AK001586.1
-
3
318
AK297193.1
-
3
296
AK298991.1
-
3
318
AK302955.1
-
2
183
AK316190.1
-
2
184
AK316524.1
-
3
411
AL049848.1
-
1
31
BC039071.1
-
3
306
5177926
BC117130.1
-
2
153
40125681
DA336461.1
-
3
350
DB077466.1
-
3
288
DB080645.1
-
3
298
Back To Top
More Information
Panther Classification:
Molecular Function -
acyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
smoothened signaling pathway
multicellular organism development
protein palmitoylation
positive regulation of protein targeting to mitochondrion
GTP binding
O-acyltransferase activity
palmitoyltransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-8tsm6:80/100.66.77.47:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline