Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PQLC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099777.3 | NP_001093247.1 | 5 | 1046 | ||
NM_001243729.1 | NP_001230658.1 | 4 | 957 | |||
XM_017005754.1 | XP_016861243.1 | 6 | 1096 |