Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286772.1 | NP_001273701.1 | 5 | 430 | ||
NM_018201.4 | NP_060671.3 | 5 | 430 | |||
XM_005252060.2 | XP_005252117.1 | 4 | 333 | |||
GenBank mRNA | AB209565.1 | - | 2 | 149 | ||
AB449899.1 | - | 5 | 283 | |||
AK001605.1 | - | 5 | 288 | |||
AK289777.1 | - | 5 | 342 | |||
AK295183.1 | - | 2 | 133 | |||
AK307556.1 | - | 1 | 32 | |||
BC014898.1 | - | 5 | 305 | |||
BC132817.1 | - | 5 | 303 | 40146839 | ||
BC136509.1 | - | 5 | 303 | 9020499 | ||
BC150311.1 | - | 5 | 339 | 8860447 | ||
DC325060.1 | - | 5 | 406 |