Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPINK13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040129.2 | NP_001035218.1 | 5 | 496 | ||
GenBank mRNA | BC128163.1 | - | 6 | 648 | 40113027 | |
BC128164.1 | - | 5 | 496 | 40113028 | ||
EF108324.1 | - | 5 | 544 | |||
EF426754.1 | - | 5 | 544 |