Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAVER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018211.3 | NP_060681.2 | 8 | 1424 | ||
XM_006710738.3 | XP_006710801.2 | 8 | 1570 | |||
XM_011541706.2 | XP_011540008.1 | 8 | 1579 | |||
GenBank mRNA | AB046799.1 | - | 8 | 1424 | ||
AK001632.1 | - | 6 | 587 | |||
AL359613.1 | - | 1 | 30 | |||
BC065303.1 | - | 6 | 854 |