Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008493.1 | NP_001008493.1 | 8 | 1707 | ||
NM_018212.4 | NP_060682.2 | 8 | 1707 | |||
XM_005273187.2 | XP_005273244.1 | 9 | 1747 | |||
XM_011544229.2 | XP_011542531.2 | 9 | 2223 | |||
XM_017001746.1 | XP_016857235.1 | 10 | 2982 | |||
XM_017001747.1 | XP_016857236.1 | 10 | 2235 | |||
XM_017001748.1 | XP_016857237.1 | 8 | 2166 | |||
XM_017001749.1 | XP_016857238.1 | 9 | 2223 | |||
XM_017001750.1 | XP_016857239.1 | 9 | 2124 | |||
XM_017001751.1 | XP_016857240.1 | 8 | 2055 | |||
XM_017001752.1 | XP_016857241.1 | 7 | 2055 | |||
XM_017001753.1 | XP_016857242.1 | 11 | 1786 | |||
XM_017001754.1 | XP_016857243.1 | 10 | 1730 | |||
XM_017001755.1 | XP_016857244.1 | 10 | 1473 | |||
XM_017001756.1 | XP_016857245.1 | 8 | 1404 | |||
XM_017001757.1 | XP_016857246.1 | 8 | 1307 | |||
GenBank mRNA | AF519769.1 | - | 8 | 1523 | ||
AK001635.1 | - | 2 | 187 | |||
AK096246.1 | - | 6 | 1003 | |||
AK126894.1 | - | 3 | 464 | |||
AK127561.1 | - | 2 | 838 | |||
AK290179.1 | - | 8 | 1651 | |||
AL133059.1 | - | 2 | 78 | |||
AY345143.1 | - | 8 | 1611 | |||
BC065238.1 | - | 7 | 1125 | |||
BC095481.1 | - | 8 | 1706 | 30348815 | ||
EU255274.1 | - | 6 | 1147 | |||
KM214203.1 | - | 8 | 1311 | |||
KM214204.1 | - | 8 | 1311 |