Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPF3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017029737.1 | XP_016885226.1 | 13 | 1788 | ||
XM_017029738.1 | XP_016885227.1 | 13 | 1788 | |||
XM_017029739.1 | XP_016885228.1 | 12 | 1749 | |||
XM_017029740.1 | XP_016885229.1 | 13 | 1686 |