Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPF3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017029737.1 | XP_016885226.1 | 14 | 1890 | ||
XM_017029738.1 | XP_016885227.1 | 14 | 1890 | |||
XM_017029739.1 | XP_016885228.1 | 13 | 1851 | |||
XM_017029740.1 | XP_016885229.1 | 14 | 1788 |