Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105669.2 | NP_001099139.2 | 2 | 648 | ||
XM_011509253.2 | XP_011507555.1 | 1 | 30 | |||
XM_011509254.2 | XP_011507556.1 | 1 | 30 | |||
XM_011509255.2 | XP_011507557.1 | 1 | 30 |