Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133475.1 | NP_597732.1 | 2 | 298 | ||
XM_011527756.2 | XP_011526058.1 | 2 | 181 | |||
XM_011527757.2 | XP_011526059.1 | 2 | 456 | |||
XM_011527758.2 | XP_011526060.1 | 2 | 181 | |||
XM_011527761.1 | XP_011526063.1 | 3 | 251 | |||
XM_011527764.2 | XP_011526066.1 | 2 | 456 | |||
XM_017026438.1 | XP_016881927.1 | 3 | 242 | |||
XM_017026439.1 | XP_016881928.1 | 3 | 251 | |||
XM_017026440.1 | XP_016881929.1 | 3 | 251 | |||
GenBank mRNA | BC028236.1 | - | 1 | 27 | ||
CN286920.1 | - | 2 | 298 |