Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133475.1 | NP_597732.1 | 6 | 665 | ||
XM_011527756.2 | XP_011526058.1 | 6 | 548 | |||
XM_011527757.2 | XP_011526059.1 | 6 | 823 | |||
XM_011527758.2 | XP_011526060.1 | 5 | 461 | |||
XM_011527761.1 | XP_011526063.1 | 7 | 618 | |||
XM_011527764.2 | XP_011526066.1 | 5 | 736 | |||
XM_011527765.2 | XP_011526067.1 | 4 | 362 | |||
XM_017026438.1 | XP_016881927.1 | 7 | 609 | |||
XM_017026439.1 | XP_016881928.1 | 6 | 531 | |||
XM_017026440.1 | XP_016881929.1 | 6 | 531 | |||
GenBank mRNA | AB075861.1 | - | 4 | 362 | ||
BC028236.1 | - | 4 | 307 | |||
BI914435.1 | - | 4 | 390 |