Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PATL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145112.1 | NP_001138584.1 | 8 | 815 | ||
XM_011521336.2 | XP_011519638.2 | 8 | 1641 | |||
XM_011521337.2 | XP_011519639.2 | 8 | 1776 | |||
XM_011521338.2 | XP_011519640.1 | 10 | 1340 | |||
XM_011521339.2 | XP_011519641.1 | 9 | 1221 | |||
XM_011521340.2 | XP_011519642.1 | 8 | 1162 | |||
XM_011521341.1 | XP_011519643.1 | 8 | 925 | |||
XM_011521342.2 | XP_011519644.1 | 6 | 1822 | |||
XM_011521343.2 | XP_011519645.1 | 9 | 1172 | |||
XM_011521344.2 | XP_011519646.1 | 6 | 614 | |||
XM_011521345.2 | XP_011519647.1 | 9 | 1134 | |||
XM_011521346.2 | XP_011519648.2 | 4 | 1348 | |||
XM_011521347.1 | XP_011519649.1 | 7 | 730 | |||
XM_011521348.2 | XP_011519650.1 | 7 | 730 | |||
XM_017022000.1 | XP_016877489.1 | 8 | 1633 | |||
XM_017022001.1 | XP_016877490.1 | 6 | 762 | |||
GenBank mRNA | BI835136.1 | - | 7 | 730 |