Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145154.1 | NP_001138626.1 | 10 | 1282 | ||
NM_001373.1 | NP_001364.1 | 10 | 1282 | |||
XM_006711735.3 | XP_006711798.1 | 10 | 2513 | |||
XM_011544057.2 | XP_011542359.1 | 10 | 2513 | |||
XM_011544058.2 | XP_011542360.1 | 10 | 2513 | |||
XM_011544059.2 | XP_011542361.1 | 10 | 2513 | |||
XM_011544061.2 | XP_011542363.1 | 10 | 2513 | |||
XM_011544062.2 | XP_011542364.1 | 10 | 2513 | |||
XM_011544063.2 | XP_011542365.1 | 10 | 2513 | |||
XM_011544064.2 | XP_011542366.1 | 10 | 2513 | |||
XM_011544065.2 | XP_011542367.1 | 10 | 2513 | |||
XM_011544066.2 | XP_011542368.1 | 10 | 2513 | |||
XM_011544067.2 | XP_011542369.1 | 10 | 2513 | |||
XM_011544069.2 | XP_011542371.1 | 10 | 2513 | |||
XM_011544070.2 | XP_011542372.1 | 10 | 2513 | |||
XM_011544072.2 | XP_011542374.1 | 10 | 2513 | |||
XM_017000295.1 | XP_016855784.1 | 10 | 2513 | |||
XM_017000296.1 | XP_016855785.1 | 10 | 2513 | |||
XM_017000297.1 | XP_016855786.1 | 10 | 2513 | |||
XM_017000298.1 | XP_016855787.1 | 10 | 2512 | |||
XM_017000300.1 | XP_016855789.1 | 10 | 2507 | |||
GenBank mRNA | BC042869.1 | - | 10 | 1184 | ||
BC119716.1 | - | 10 | 1269 | |||
BC119717.1 | - | 10 | 1269 |