Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PABPC1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134984.1 | 1 | 572 | |||
NR_134985.1 | 1 | 572 | ||||
NR_134986.1 | 1 | 572 | ||||
NR_134987.1 | 1 | 572 | ||||
GenBank mRNA | AK124047.1 | - | 1 | 572 | ||
CB152000.1 | - | 1 | 468 |