Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TET3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287491.1 | NP_001274420.1 | 11 | 4650 | ||
XM_005264187.3 | XP_005264244.1 | 12 | 5170 | |||
XM_011532682.2 | XP_011530984.1 | 13 | 5218 | |||
XM_011532683.2 | XP_011530985.1 | 13 | 5527 | |||
XM_011532684.2 | XP_011530986.1 | 13 | 6959 | |||
XM_011532685.2 | XP_011530987.1 | 12 | 5161 | |||
XM_011532686.2 | XP_011530988.1 | 11 | 5113 | |||
XM_011532687.2 | XP_011530989.1 | 12 | 4500 | |||
XM_011532689.2 | XP_011530991.1 | 11 | 2279 | |||
XM_011532690.1 | XP_011530992.1 | 10 | 2336 | |||
XM_017003566.1 | XP_016859055.1 | 13 | 7058 | |||
XM_017003567.1 | XP_016859056.1 | 11 | 4482 | |||
GenBank mRNA | AB007861.1 | - | 1 | 647 | ||
BC022243.1 | - | 8 | 2106 | 4400878 | ||
HQ220209.1 | - | 11 | 4650 |