Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308384.1 | NP_001295313.1 | 3, 4 | 1338 | ||
NM_018231.2 | NP_060701.1 | 4, 5 | 870 | |||
XM_017023393.1 | XP_016878882.1 | 3, 4 | 1351 | |||
XM_017023394.1 | XP_016878883.1 | 4, 5 | 846 | |||
XM_017023395.1 | XP_016878884.1 | 4, 5 | 846 | |||
XM_017023396.1 | XP_016878885.1 | 4, 5 | 846 | |||
XM_017023397.1 | XP_016878886.1 | 4, 5 | 846 | |||
XM_017023398.1 | XP_016878887.1 | 4, 5 | 846 | |||
XM_017023399.1 | XP_016878888.1 | 4, 5 | 846 | |||
GenBank mRNA | AK001677.1 | - | 4, 5 | 863 | ||
AK022786.1 | - | 4, 5 | 836 | |||
AK222932.1 | - | 4, 5 | 832 | |||
AK299424.1 | - | 4, 5 | 495 | |||
BC001961.1 | - | 3, 4 | 819 | 3510557 | ||
BC063399.1 | - | 3, 4 | 1338 | |||
CR457236.1 | - | 2, 3 | 470 |