Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: R3HCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136108.2 | NP_001129580.2 | 4 | 662 | ||
NM_001301650.1 | NP_001288579.1 | 5 | 536 | |||
NR_125897.1 | 4 | 662 | ||||
GenBank mRNA | AK293465.1 | - | 4 | 646 | ||
AL050297.1 | - | 4 | 640 | |||
BC032421.1 | - | 4 | 629 | |||
BC050572.1 | - | 3 | 601 | |||
BC143822.1 | - | 5 | 720 | 9052337 | ||
BC143827.1 | - | 4 | 846 | |||
BC143829.1 | - | 4 | 846 | 9052344 | ||
BX426634.2 | - | 4 | 675 |