Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM218 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080546.2 | NP_001074015.1 | 5 | 1227 | ||
NM_001258238.1 | NP_001245167.1 | 5 | 1105 | |||
NM_001258239.1 | NP_001245168.1 | 5 | 1202 | |||
NM_001258240.1 | NP_001245169.1 | 7 | 1265 | |||
NM_001258241.1 | NP_001245170.1 | 6 | 1206 | |||
NM_001258242.1 | NP_001245171.1 | 5 | 1193 | |||
NM_001258243.1 | NP_001245172.1 | 6 | 1296 | |||
NM_001258244.1 | NP_001245173.1 | 5 | 1096 | |||
NM_001258245.1 | NP_001245174.1 | 7 | 1258 | |||
NM_001258246.1 | NP_001245175.1 | 5 | 1178 | |||
NM_001258247.1 | NP_001245176.1 | 5 | 1081 | |||
NR_047586.1 | 5 | 1058 | ||||
NR_047587.1 | 3 | 841 | ||||
NR_047588.1 | 5 | 1146 | ||||
NR_047589.1 | 4 | 910 | ||||
NR_047590.1 | 5 | 1083 | ||||
NR_047591.1 | 5 | 969 | ||||
NR_047592.1 | 5 | 1049 | ||||
XM_006718779.3 | XP_006718842.1 | 7 | 1406 | |||
XM_006718780.3 | XP_006718843.1 | 6 | 1303 | |||
XM_006718781.3 | XP_006718844.1 | 7 | 1360 | |||
XM_006718783.3 | XP_006718846.1 | 5 | 1169 | |||
XM_017017322.1 | XP_016872811.1 | 6 | 1193 | |||
XM_017017323.1 | XP_016872812.1 | 5 | 1119 | |||
XM_017017324.1 | XP_016872813.1 | 5 | 1121 | |||
GenBank mRNA | AK091324.1 | - | 2 | 4404 | ||
AL833215.1 | - | 2 | 808 | |||
CX870447.1 | - | 1 | 222 |