Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMEM218 SIRNA Assays ›
Gene Symbol:
TMEM218
Gene Name:
transmembrane protein 218
Gene Aliases:
-
Chromosome Location:
Chr.11: 125094370 - 125111763 on Build GRCh38
UniGene ID:
Hs.731899
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
264626
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMEM218
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080546.2
NP_001074015.1
5
776
NM_001258238.1
NP_001245167.1
5
654
NM_001258239.1
NP_001245168.1
5
751
NM_001258240.1
NP_001245169.1
7
814
NM_001258241.1
NP_001245170.1
6
755
NM_001258242.1
NP_001245171.1
5
742
NM_001258243.1
NP_001245172.1
6
845
NM_001258244.1
NP_001245173.1
5
645
NM_001258245.1
NP_001245174.1
7
807
NM_001258246.1
NP_001245175.1
5
727
NM_001258247.1
NP_001245176.1
5
630
NR_047586.1
5
607
NR_047587.1
3
390
NR_047588.1
5
695
NR_047589.1
4
459
NR_047590.1
5
632
NR_047591.1
5
518
NR_047592.1
5
598
XM_006718779.3
XP_006718842.1
7
955
XM_006718780.3
XP_006718843.1
6
852
XM_006718781.3
XP_006718844.1
7
909
XM_006718783.3
XP_006718846.1
5
718
XM_017017322.1
XP_016872811.1
6
742
XM_017017323.1
XP_016872812.1
5
668
XM_017017324.1
XP_016872813.1
5
670
GenBank mRNA
AI680968.1
-
1
84
AK091324.1
-
2
3953
AL833215.1
-
2
357
BC132716.1
-
5
613
40146738
BC132718.1
-
5
613
40146740
BC144270.1
-
6
716
9052790
BC144276.1
-
5
566
BC144277.1
-
5
647
9052797
BE513148.1
-
5
586
BE974033.1
-
3
235
BI192743.1
-
5
499
BI547955.1
-
5
404
BI829527.1
-
5
589
BI856825.1
-
5
520
BM042315.1
-
6
606
BQ223298.1
-
7
702
BQ225039.1
-
4
376
BQ420663.1
-
5
621
BQ653243.1
-
5
621
BQ681518.1
-
6
604
BU160644.1
-
5
451
BU598207.1
-
5
548
BU855096.1
-
5
542
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-8skhh:80/10.239.164.194:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline