Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018117.11 | NP_060587.8 | 16 | 2337 | ||
XM_005269962.2 | XP_005270019.1 | 16 | 2054 | |||
XM_005269963.2 | XP_005270020.1 | 16 | 2186 | |||
XM_005269964.2 | XP_005270021.1 | 9 | 1092 | |||
XM_017016397.1 | XP_016871886.1 | 15 | 2025 | |||
XM_017016398.1 | XP_016871887.1 | 15 | 1955 | |||
XM_017016399.1 | XP_016871888.1 | 15 | 1899 | |||
XM_017016400.1 | XP_016871889.1 | 16 | 2228 | |||
GenBank mRNA | AB037772.2 | - | 16 | 2150 | ||
AF320223.1 | - | 16 | 2124 | |||
AK000579.1 | - | 2 | 139 | |||
AK001368.1 | - | 11 | 1240 | |||
AK299524.1 | - | 9 | 957 | |||
AK315145.1 | - | 16 | 2115 | |||
AL137699.1 | - | 14 | 1810 | |||
BC040469.1 | - | 16 | 2154 | 4730308 | ||
BC071564.1 | - | 16 | 2120 | 30346203 |