Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWDE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135924.1 | NP_001129396.1 | 21 | 4511 | ||
XM_005249641.1 | XP_005249698.1 | 18 | 3625 | |||
XM_011515174.1 | XP_011513476.1 | 23 | 4548 | |||
XM_011515175.1 | XP_011513477.1 | 22 | 4410 | |||
XM_017011828.1 | XP_016867317.1 | 19 | 3843 | |||
XM_017011829.1 | XP_016867318.1 | 18 | 3512 | |||
GenBank mRNA | AK027618.1 | - | 10 | 2098 | ||
BC144316.1 | - | 22 | 4895 | |||
BC144319.1 | - | 18 | 3663 | 9052839 |