Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFR3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319099.1 | NP_001306028.1 | 7 | 955 | ||
NM_014971.1 | NP_055786.1 | 7 | 967 | |||
XM_011532701.2 | XP_011531003.1 | 7 | 967 | |||
GenBank mRNA | AB023170.1 | - | 7 | 941 | ||
AK095190.1 | - | 7 | 955 | |||
BC049384.1 | - | 4 | 511 | 5742159 |