Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC39A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080494.3 | NP_001073963.1 | 16, 17 | 1599 | ||
NM_001144832.2 | NP_001138304.1 | 16, 17 | 1574 | |||
NM_001297663.1 | NP_001284592.1 | 16, 17 | 1576 | |||
NM_001297664.1 | NP_001284593.1 | 16, 17 | 1612 | |||
NM_001297665.1 | NP_001284594.1 | 16, 17 | 1704 | |||
NM_001297666.1 | NP_001284595.1 | 5, 6 | 607 | |||
NM_001297667.1 | NP_001284596.1 | 4, 5 | 515 | |||
XM_005270644.2 | XP_005270701.1 | 16, 17 | 1605 | |||
XM_006710471.2 | XP_006710534.1 | 16, 17 | 1702 | |||
XM_011541048.2 | XP_011539350.1 | 16, 17 | 1679 | |||
XM_011541049.2 | XP_011539351.1 | 16, 17 | 1818 | |||
XM_011541050.2 | XP_011539352.1 | 16, 17 | 1679 | |||
XM_011541051.2 | XP_011539353.1 | 16, 17 | 1712 | |||
XM_011541052.2 | XP_011539354.1 | 16, 17 | 1658 | |||
XM_011541053.2 | XP_011539355.1 | 16, 17 | 1696 | |||
XM_011541054.2 | XP_011539356.1 | 16, 17 | 1532 | |||
XM_017000715.1 | XP_016856204.1 | 16, 17 | 1579 | |||
GenBank mRNA | AF007170.1 | - | 16, 17 | 1480 | ||
AK298019.1 | - | 16, 17 | 1583 | |||
AK301587.1 | - | 16, 17 | 1572 | |||
BC004399.1 | - | 5, 6 | 568 | |||
BC028374.2 | - | 16, 17 | 1646 | |||
BQ227134.1 | - | 4, 5 | 526 |