Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC39A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080494.3 | NP_001073963.1 | 18 | 2003 | ||
NM_001144832.2 | NP_001138304.1 | 18 | 1978 | |||
NM_001297663.1 | NP_001284592.1 | 18 | 1980 | |||
NM_001297664.1 | NP_001284593.1 | 18 | 2016 | |||
NM_001297665.1 | NP_001284594.1 | 18 | 2108 | |||
NM_001297666.1 | NP_001284595.1 | 7 | 1011 | |||
NM_001297667.1 | NP_001284596.1 | 6 | 919 | |||
XM_005270644.2 | XP_005270701.1 | 18 | 2009 | |||
XM_006710471.2 | XP_006710534.1 | 18 | 2106 | |||
XM_011541048.2 | XP_011539350.1 | 18 | 2083 | |||
XM_011541049.2 | XP_011539351.1 | 18 | 2222 | |||
XM_011541050.2 | XP_011539352.1 | 18 | 2083 | |||
XM_011541051.2 | XP_011539353.1 | 18 | 2116 | |||
XM_011541052.2 | XP_011539354.1 | 18 | 2062 | |||
XM_011541053.2 | XP_011539355.1 | 18 | 2100 | |||
XM_011541054.2 | XP_011539356.1 | 18 | 1936 | |||
XM_017000715.1 | XP_016856204.1 | 18 | 1983 | |||
GenBank mRNA | AF007170.1 | - | 18 | 1884 | ||
AK298019.1 | - | 18 | 1987 | |||
AK301587.1 | - | 18 | 1976 | |||
BC004399.1 | - | 7 | 972 | |||
BC028374.2 | - | 18 | 2050 |