Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC39A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080494.3 | NP_001073963.1 | 1 | 127 | ||
NM_001297665.1 | NP_001284594.1 | 1 | 127 | |||
XM_011541049.2 | XP_011539351.1 | 1 | 122 | |||
GenBank mRNA | BC028374.2 | - | 1 | 69 | ||
DB460455.1 | - | 1 | 125 | |||
HY029911.1 | - | 1 | 128 |