Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF292 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015021.1 | NP_055836.1 | 8 | 4707 | ||
XM_005248697.4 | XP_005248754.1 | 8 | 4521 | |||
XM_011535625.2 | XP_011533927.1 | 7 | 4527 | |||
XM_017010575.1 | XP_016866064.1 | 9 | 7898 | |||
XM_017010576.1 | XP_016866065.1 | 10 | 5424 | |||
XM_017010577.1 | XP_016866066.1 | 8 | 7753 | |||
XM_017010578.1 | XP_016866067.1 | 10 | 10406 | |||
XM_017010579.1 | XP_016866068.1 | 10 | 7432 | |||
XM_017010580.1 | XP_016866069.1 | 10 | 4853 | |||
XM_017010581.1 | XP_016866070.1 | 9 | 4839 | |||
XM_017010582.1 | XP_016866071.1 | 8 | 7549 | |||
XM_017010583.1 | XP_016866072.1 | 9 | 5146 | |||
XM_017010584.1 | XP_016866073.1 | 8 | 9522 | |||
GenBank mRNA | AB011102.1 | - | 1 | 1187 | ||
AK023626.1 | - | 1 | 2408 | |||
AK127835.1 | - | 1 | 3459 | |||
AL832366.1 | - | 1 | 2977 | |||
BC016386.1 | - | 1 | 583 | |||
BX537746.1 | - | 8 | 4681 |