Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAND2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162499.1 | NP_001155971.1 | 7 | 930 | ||
NM_012298.2 | NP_036430.1 | 5 | 651 | |||
XM_011533503.2 | XP_011531805.1 | 7 | 948 | |||
XM_011533504.2 | XP_011531806.1 | 7 | 1044 | |||
GenBank mRNA | AB014567.1 | - | 5 | 579 | ||
BC136592.1 | - | 7 | 869 | 9020582 | ||
DB471722.1 | - | 3 | 368 | |||
GU727621.1 | - | 7 | 930 |