Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETD1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015048.1 | NP_055863.1 | 17 | 6563 | ||
XM_005253858.4 | XP_005253915.1 | 17 | 6851 | |||
XM_006719296.3 | XP_006719359.1 | 17 | 6767 | |||
GenBank mRNA | AB028999.1 | - | 7 | 3201 | ||
BC047909.1 | - | 1 | 482 | |||
BC080192.1 | - | 1 | 730 |