Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098811.1 | NP_001092281.1 | 10 | 1784 | ||
XM_005271927.2 | XP_005271984.1 | 10 | 1783 | |||
XM_005271928.2 | XP_005271985.1 | 10 | 1780 | |||
XM_005271931.3 | XP_005271988.1 | 11 | 1867 | |||
XM_011543288.1 | XP_011541590.1 | 10 | 1806 | |||
XM_017009254.1 | XP_016864743.1 | 10 | 1777 | |||
XM_017009255.1 | XP_016864744.1 | 10 | 1850 | |||
XM_017009256.1 | XP_016864745.1 | 10 | 1847 | |||
XM_017009257.1 | XP_016864746.1 | 10 | 1800 | |||
GenBank mRNA | AF440762.1 | - | 10 | 1784 |