Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015168.1 | NP_055983.1 | 3 | 230 | ||
XM_005258676.3 | XP_005258733.1 | 2 | 1505 | |||
XM_005258677.4 | XP_005258734.1 | 3 | 356 | |||
XM_005258678.2 | XP_005258735.1 | 2 | 113 | |||
XM_006723113.3 | XP_006723176.1 | 3 | 1920 | |||
XM_011526668.2 | XP_011524970.1 | 3 | 132 | |||
XM_011526669.2 | XP_011524971.1 | 2 | 260 | |||
XM_011526670.2 | XP_011524972.1 | 2 | 856 | |||
XM_017026529.1 | XP_016882018.1 | 2 | 1312 | |||
XM_017026530.1 | XP_016882019.1 | 3 | 1920 | |||
XM_017026531.1 | XP_016882020.1 | 2 | 166 | |||
GenBank mRNA | AB028987.2 | - | 3 | 230 | ||
AK125279.1 | - | 2 | 113 |