Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300727.1 | NP_001287656.1 | 6 | 1182 | ||
NM_015177.1 | NP_055992.1 | 6 | 1591 | |||
XM_006718483.3 | XP_006718546.1 | 6 | 1635 | |||
GenBank mRNA | AB023154.1 | - | 6 | 1438 | ||
BC119011.1 | - | 6 | 1182 | 40085251 | ||
BC122861.1 | - | 6 | 1182 | 40085252 |