Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOC6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321729.1 | NP_001308658.1 | 23 | 4161 | ||
NM_001321730.1 | NP_001308659.1 | 22 | 4026 | |||
NM_001321731.1 | NP_001308660.1 | 23 | 4202 | |||
NM_001321733.1 | NP_001308662.1 | 21 | 4014 | |||
NM_001321734.1 | NP_001308663.1 | 22 | 3995 | |||
NM_015189.2 | NP_056004.1 | 22 | 4149 | |||
NR_135773.1 | 22 | 4087 | ||||
NR_135774.1 | 20 | 3827 | ||||
XM_005264224.1 | XP_005264281.1 | 16 | 3453 | |||
XM_017003641.1 | XP_016859130.1 | 19 | 3595 | |||
XM_017003642.1 | XP_016859131.1 | 17 | 3208 | |||
GenBank mRNA | AK023791.1 | - | 10 | 2710 |